Order Entry
Norway
Orders LinkContactUsLinkComponent
Thymidine (2'-deoxythymidine)

Thymidine (2'-deoxythymidine)

Health
Irritant
Supplier:   RESEARCH ORGANICS
Thymidine (2'-deoxythymidine)
Supplier:  RESEARCH ORGANICS
Certificates
Thymidine (2'-deoxythymidine)

2 Options Available Below

Product Details & Documents
Synonyms:  1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione, 1-(2-Deoxy-β-D-ribofuranosyl)thymine, 1-(2-Deoxy-β-D-ribofuranosyl)-5-methyluracil, dT, Thymine deoxyriboside, 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione, Pyrimidine deoxynucleoside, Thymidine kinase, Thymidine-2-deoxythymidine
  • CAS Number:50-89-5
  • Formula:C₁₀H₁₄N₂O₅
  • MDL Number:MFCD00006537
  • Molecular Weight:242.23 g/mol
  •  :M-Clarity™ quality level = MQ300
  • Storage Temperature:Ambient
  • EINECS:200-070-4
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g