Guanosine-5'-monophosphate disodium salt
Catalog # MOLE66211644-5G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:407.187 g/mol
Storage Temperature:Freezer
EINECS:226-914-1
CAS Number:5550-12-9
:MFCD00070575
Formula:C₁₀H₁₂N₅Na₂O₈P
MDL Number:MFCD00151238