
Naphthalene black 10B (naphthol blue black, acid black 1) 80% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:1064-48-8
:
Formula:C₂₂H₁₄N₆Na₂O₉S₂
MDL Number:MFCD00004017
:616.49 g/mol
:M-Clarity™ quality level = MQ200
EINECS:213-903-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance (Color):Brown to Very Dark Brown and Black
- Appearance (Form):Powder
- Infrared spectrum:Conforms to Structure
- Wavelength c = 0.007g/l in H2O:616 - 622 nm
- Extinction Coefficient:≥42000
- Wavelength:318 - 324 nm
- TLC Consistent With Control:Conforms
- Solubility (Turbidity) Opaque (c = 1mg/ml; H2O):Conforms
- Solubility (Color):Dark Blue to Very Dark Blue
- Water (by Karl Fischer):≤10.0 %
- Carbon:≥33.4 %
- Nitrogen:≥10.6 %