N,N'-Diphenylbenzidine for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:531-91-9
:
Formula:C₂₄H₂₀N₂
MDL Number:MFCD00003016
:336.44 g/mol
:M-Clarity™ quality level = MQ200
EINECS:208-521-7
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:245 - 250 °C
- Molar Mass:336.43 g/mol
- Synonyms:N,N'-Diphenyl-4,4'-diaminobiphenyl, N,N'-Diphenyl-[1,1'-biphenyl]-4,4'-diamine
- Hill Formula:C24H20N2
- Chemical Formula:4-(C6H5NH)C6H4C6H4-4-(NHC6H5)