N,N'-Diphenylbenzidine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:531-91-9
:
Formula:C₂₄H₂₀N₂
MDL Number:MFCD00003016
:336.44 g/mol
Merck Index:14,3319
Melting Pt:246…250 °C
Storage Temperature:Ambient
EINECS:208-521-7