Order Entry
Norway
Orders LinkContactUsLinkComponent
Tris acetate

Tris acetate

Glass bottle for solids, 1 kg
Catalog # RESERES3090T-A103X
Irritant
Supplier:   RESEARCH ORGANICS
Tris acetate
Catalog # RESERES3090T-A103X
Supplier:  RESEARCH ORGANICS
CAS Number:  6850-28-8
Tris acetate

Glass bottle for solids, 1 kg

CAS Number:  6850-28-8
Price
Inquire For Price
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Tris(hydroxymethyl)aminomethane acetate salt, 2-Amino-2-(hydroxymethyl)propane-1,3-diol acetate salt, Trimethylolaminomethane acetate, Tris acetate, Tris(hydroxymethyl)aminomethane acetate, 2-Amino-2-(hydroxymethyl)-1,3-propanediol acetate
  • CAS Number:6850-28-8
  • Formula:NH₂C(CH₂OH)₃·CH₃COOH
  • MDL Number:MFCD00038951
  • Molecular Weight:181.1870 g/mol
  • Storage Temperature:Ambient
  • EINECS:229-939-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.