Creatine phosphate disodium salt, hydrate ≥98%, powder
Catalog # ICNA0219923505
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Identity Test:Passes
- Purity:≥98%
- FTIR:Conforms to Standard
- pH (1% aq soln):7 - 9
- Moisture:22±3%
- Transmittance:99.10%
Specification Test Results
About this item
:
:255.077 g/mol
EINECS:213-074-6
CAS Number:922-32-7
:19333-65-4
Formula:C₄H₈N₃O₅P·2Na
MDL Number:MFCD00014351