Order Entry
Norway
ContactUsLinkComponent
o-Cresolphthalein complexone indicator for complexometry, Fluka™
o-Cresolphthalein complexone indicator for complexometry, Fluka™
Certificates
o-Cresolphthalein complexone indicator for complexometry, Fluka™
Specification Test Results
  • Ignition Residue:
    ~1%
  • Appearance of the substance:
    complying
  • Identity (LC-MS):
    complying
  • Indicator solution:
    complying
  • Water (Karl Fischer):
    Max. 10 %
  • Residue on ignition:
    Max. 2 %
  • Suitability for Complexometry:
    complying