Order Entry
Norway
ContactUsLinkComponent
o-Cresolphthalein complexone Reag. Ph. Eur. indicator for metal titration, Supelco®
o-Cresolphthalein complexone Reag. Ph. Eur. indicator for metal titration, Supelco®
Supplier:  Merck
Certificates
o-Cresolphthalein complexone Reag. Ph. Eur. indicator for metal titration, Supelco®
Supplier:  Merck
Specification Test Results
  • Identity (UV/VIS-Spectrum):
    passes test
  • Absorption maximum λmax. (Sodium hydroxide solution 0.1 mol/l):
    577 - 580 nm
  • Spec. Absorptivity A 1%/1cm (λmax; 0.005 g/l; sodium hydroxide solution 0.1 mol/l; calc. on anhydrous substance):
    ≥ 750
  • Water (according to Karl Fischer):
    ≤ 10 %
  • Suitability as indicator (for metal titration):
    passes test