

Big CHAP (N,N-Bis[3-(D-gluconamido)propyl]cholamide), Millipore®
Catalog # 200965-1
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:GHS06
:GHS08
Storage Temperature:Refrigerator
CAS Number:86303-22-2
Formula:C₄₂H₇₅N₃O₁₆
MDL Number:MFCD00082541