Order Entry
Norway
ContactUsLinkComponent
Azure B (azure I)
Azure B (azure I)
Catalog # CHRO1B-487
Supplier:  CHROMA
CAS Number:  
Azure B (azure I)
Catalog # CHRO1B-487
Supplier:  CHROMA
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :305.83 g/mol
Storage Temperature:Ambient
EINECS:208-511-2
CAS Number:531-55-5
Formula:C₁₅H₁₆ClN₃S
MDL Number:MFCD00011935