Order Entry
Norway
ContactUsLinkComponent
Streptavidin
Streptavidin
Supplier:  PanReac AppliChem
Certificates
Streptavidin
Supplier:  PanReac AppliChem

Specifications

  • CAS No.:
    9013-20-1
  • Storage conditions:
    Freezer
  • MDL No.:
    MFCD00082035
  • Protein/peptide name:
    Streptavidin

Specifications