Order Entry
Norway
ContactUsLinkComponent
Cresol red, pure indicator grade
Cresol red, pure indicator grade
Certificates
undefined
Cresol red, pure indicator grade
Certificates

Specification Test Results

  • Appearance:
    Dark green to red-brown Powder
  • Infrared spectrum:
    Conforms
  • Solubility:
    (0.1 % in 50 % ethanol) Clear
  • Specific extinction E (1%/1cm):
    ≥1500 (572 nm, in 0.01 N NaOH)
  • Visual transition interval:
    from pH 7.0 (orange) to pH 8.8 (purple)

Specification Test Results

Related Information