Order Entry
Norway
ContactUsLinkComponent
Hafnium standard solution, 1,000 mg/L Hf in 2% hydrochloric acid (from HfOCl2) standard for AAS and ICP
Hafnium standard solution, 1,000 mg/L Hf in 2% hydrochloric acid (from HfOCl2) standard for AAS and ICP
Catalog # PERCN9300122
Leverandør:  PerkinElmer
CAS-nummer:  
Hafnium standard solution, 1,000 mg/L Hf in 2% hydrochloric acid (from HfOCl2) standard for AAS and ICP
Catalog # PERCN9300122
Leverandør:  PerkinElmer
CAS-nummer:  

Spesifikasjoner

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HCl
  • Pk:
    1
  • Pk Info:
    1×500 ml
  • Element:
    Hafnium
  • Symbol:
    Hf

Spesifikasjoner

Om dette produktet

Pure Grade standard for AA and ICP-OES

  • Analysed by ICP-OES
  • Analysed by Classical Wet Assay
  • 32 trace impurities analyzed by ICP-MS of the final solution and reported on the certificate
  • Impurities reported at ppm level
  • All Standards are prepared and certified under ISO Guide 34 and ISO 17025 – certified by A2LA