Order Entry
Norway
ContactUsLinkComponent
Fixation plasters, Askina® silk
Fixation plasters, Askina® silk
  BRAU9080120N
 :  B. Braun
undefined
Fixation plasters, Askina® silk
  BRAU9080120N
 :  B. Braun
  • Description:
    Fixation plaster, for all types of wound dressings, roll of 9,1 m
  • L×W:
    9,1 m × 25 mm
  • Pk:
    12
Silk plasters for the fixation of all types of wound dressing, e.g. Askina® Pad, Askina® gauze compresses. Also suitable for the attachment of catheters, tubes and puncture sets.
  • Reliable adhesion on different surfaces
  • Skin-friendly acrylate adhesive
  • Very easy to tear, removable with little pain