Order Entry
Norway
ContactUsLinkComponent
 
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) ≥98%, Ultra Pure Grade
  0145-250MG
 :  
undefined
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) ≥98%, Ultra Pure Grade
  0145-250MG
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    250 mg
  • Seq:
    2590

 

 

  • Moisture (KF):
    ≤13%
  • Purity (Dried):
    ≥98%

 

 

 :
 :550.09 g/mol
 :Freezer
 :243-347-5
 :19817-92-6
 :C₉H₁₂N₂Na₃O₁₅P₃
 :MFCD00044310