Order Entry
Norway
ContactUsLinkComponent
5-Iodo-2'-deoxyuridine (Idoxuridine)
5-Iodo-2'-deoxyuridine (Idoxuridine)
Catalog # PROOMM0791.00
CAS Number:  
5-Iodo-2'-deoxyuridine (Idoxuridine)
Catalog # PROOMM0791.00
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :354.10 g/mol
Storage Temperature:Ambient
EINECS:200-207-8
CAS Number:54-42-2
Formula:C₉H₁₁IN₂O₅
MDL Number:MFCD00134656