Order Entry
Norway
ContactUsLinkComponent
N-Octyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-8, SB-8)
Irritant
N-Octyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-8, SB-8)
Catalog # MOLEM69597338
Supplier:  Molekula
CAS Number:  
Irritant
N-Octyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-8, SB-8)
Catalog # MOLEM69597338
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :GHS07
Storage Temperature:Ambient
CAS Number:15178-76-4
Formula:C₁₃H₂₉NO₃S
MDL Number:MFCD00036912