Order Entry
Norway
ContactUsLinkComponent
Propylene carbonate
Propylene carbonate
Catalog # EHERC16527000
Supplier:  EHRENSTORFER
CAS Number:  
Propylene carbonate
Catalog # EHERC16527000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

About this item

 :
 :102.09 g/mol
Boiling Pt:241…242 °C (1013 hPa)
Melting Pt:–48.8 °C
Density:1.2 g/cm³ (20 °C)
Flash Pt:132 °C (closed cup)
Storage Temperature:Ambient
EINECS:203-572-1
 :GHS07
CAS Number:108-32-7
Formula:C₄H₆O₃
MDL Number:MFCD00005385