Lanyard, Miller® Titan™
Supplier: HONEYWELL SAFETY
Certificates
About this item
These lanyards, consisting of an adjustable polyamide guy rope with loop and one or two twist lock carabiners, are suitable for a variety of applications, including but not limited to the construction industry and maintenance.
- Application: Work positioning
- Ideal for anyone who needs to work at heights with free hands in a secure position with sufficient hold
Certifications: EN 358
Specifications
- Catalog No:
- DALL1008287
- DALL1008288
- Material:
- Polyamide
- Polyamide
- Breaking strength:
- > 1500 daN
- > 1500 daN