Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Desoxyribonuclease I
Desoxyribonuclease I
Desoxyribonuclease I
Catalog # 69182-3
Supplier:
Merck Millipore (Novagen)
CAS Number:
Expand image
Desoxyribonuclease I
Catalog # 69182-3
Supplier:
Merck Millipore (Novagen)
CAS Number:
Specifications
Pk:
1.000 EU
CAS No.:
9003-98-9
Storage conditions:
Freezer
Enzyme name:
Desoxyribonuclease I
Enzyme synonyms:
DNase I
Specifications
About this item
Product Details
Product Details