Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Flurprimidol
Flurprimidol
Flurprimidol
Flurprimidol
Catalog # EHERC13851000
Supplier:
EHRENSTORFER
CAS Number:
Expand image
Flurprimidol
Catalog # EHERC13851000
Supplier:
EHRENSTORFER
CAS Number:
Specifications
Pack type:
Glass bottle for solids
Pk:
0,1 g
Seq:
3020
Specifications
About this item
Product Details
:
CAS Number:
56425-91-3
Formula:
C₁₅H₁₅F₃N₂O₂
Product Details