Order Entry
Norway
ContactUsLinkComponent
5-Fluoroorotic acid monohydrate (5-FOA)
5-Fluoroorotic acid monohydrate (5-FOA)
Catalog # USBIF5050-500
Supplier:  US Biological
CAS Number:  
5-Fluoroorotic acid monohydrate (5-FOA)
Catalog # USBIF5050-500
Supplier:  US Biological
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :174.0 g/mol (anhy)
EINECS:211-876-0
CAS Number:220141-70-8
 :703-95-7
Formula:C₅H₃FN₂O₄·1H₂O
MDL Number:MFCD00150658