Sulphacetamide
Katalog # EHERC16988850
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:214.24 g/mol
EINECS:205-640-6
CAS Number:144-80-9
Formula:C₈H₁₀N₂O₃S
MDL Number:MFCD00066501