Order Entry
Norway
ContactUsLinkComponent
 
5-Methylindoline ≥98.0% (by GC, titration analysis)
  TCIAM1372-5G
 :  TCI
 :  
undefined
5-Methylindoline ≥98.0% (by GC, titration analysis)
  TCIAM1372-5G
 :  TCI
 :  

 

  • Pakketype:
    Glass bottle
  • Fp:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    Colorless to Yellow to Green clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

 

 

 :
 :133.1900 g/mol
 :GHS07
 :65826-95-1
 :C₉H₁₁N
 :MFCD03093636