Order Entry
Norway
ContactUsLinkComponent
 
Organic reference standard, 4-Ethylphenol 10 µg/ml in acetonitrile, Dr Ehrenstorfer
  EHERL13352000AL
 :  
Organic reference standard, 4-Ethylphenol 10 µg/ml in acetonitrile, Dr Ehrenstorfer
  EHERL13352000AL
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    123-07-9
  • Description:
    4-Ethylphenol 10 µg/ml in acetonitrile
  • Formula:
    C₈H₁₀O
  • Description:
    4-Ethylphenol