Skip to Header
Skip to Main Content
Skip to Footer
Avansert
Avansert
SDS
Sertifikat
Produsentdata
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(Trifluoromethyl)benzyl mercaptan
3-(Trifluoromethyl)benzyl mercaptan
3-(Trifluoromethyl)benzyl mercaptan
3-(Trifluoromethyl)benzyl mercaptan
Supplier:
Thermo Scientific
Certificates
Utvid bildet
3-(Trifluoromethyl)benzyl mercaptan
Supplier:
Thermo Scientific
About this item
CAS Number:
25697-55-6
:
Formula:
C₈H₇F₃S
MDL Number:
MFCD00042435
:
192.2 g/mol
Boiling Pt:
69…70 °C (1013 hPa)
UN:
2810
ADR:
6.1,III