Butter trier
Supplier: GERBER FUNKE
Certificates
About this item
Chrome-nickel steel. Developed to the test the firmness and consistency of butter. For use in butter making industries.
- With metal handle for improved hygiene
- Two different sizes available