Bellows samplers, SP Bel-Art
BELAH378230050
- Capacity:50 ml
- Length:500 mm
- Pk:5
These bellows samplers made of PE provide quick and easy liquid sampling with one hand operation.
- FDA grade polyethylene sampler tube
- Long tapered tip with a 4 mm opening
- Easy one-hand operation
: Five per bag