Pesticide reference standard, Chlorfenvinphos (2-Chloro-1-(2,4 dichlorophenyl) vinyl diethyl phosphate, mixture of cis and trans isomers) 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERXA11290000CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:470-90-6
- Description:Chlorfenvinphos (2-Chloro-1-(2,4 dichlorophenyl) vinyl diethyl phosphate, mixture of cis and trans isomers) 100 µg/ml in cyclohexane
- Formula:C₁₂H₁₄Cl₃O₄P
- Description:Chlorfenvinphos (2-Chloro-1-(2,4 dichlorophenyl) vinyl diethyl phosphate, mixture of cis and trans isomers)