Carbaryl(1-naphthyl methylcarbamate)
Catalog # PROOBP523
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
ADR:6.1,III
:201.22 g/mol
EINECS:200-555-0
UN:2757
CAS Number:63-25-2
Formula:C₁₂H₁₁NO₂