Tantalum standard solution, 1000 mg/L Ta in 2% hydrofluoric acid (from TaCl₅), standard for AAS, Specpure®
MDL No.: MFCD00011252
Molecular Formula: TaCl₅ in 2% HF
UN No.: 2922
ADR: 8,II
GHS Symbol(s): GHS06-GHS05
GHS Signal Word: Danger
: Traceable to NIST
- MDL No.:MFCD00011252