Pesticide reference standard, 2,4,5,6-Tetrachloro-m-xylene 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA17382500ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:877-09-8
- Description:2,4,5,6-Tetrachloro-m-xylene 100 µg/ml in methanol
- Formula:C₈H₆Cl
- Description:2,4,5,6-Tetrachloro-m-xylene