Order Entry
Norway
ContactUsLinkComponent
 
Pesticide reference standard, 2,4,5,6-Tetrachloro-m-xylene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
  EHERL17382500CY
 :  
Pesticide reference standard, 2,4,5,6-Tetrachloro-m-xylene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
  EHERL17382500CY
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    877-09-8
  • Description:
    2,4,5,6-Tetrachloro-m-xylene 10 µg/ml in cyclohexane
  • Formula:
    C₈H₆Cl
  • Description:
    2,4,5,6-Tetrachloro-m-xylene