kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
3,3'-Diaminobenzidine
3,3'-Diaminobenzidine (DAB) 99%
3,3'-Diaminobenzidine (DAB) 99%
3,3'-Diaminobenzidine (DAB) 99%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
3,3'-Diaminobenzidine (DAB) 99%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
91-95-2
:
Formula:
C₁₂H₁₄N₄
MDL Number:
MFCD00007725
:
214.27 g/mol
Melting Pt:
175…177 °C
EINECS:
202-110-6
:
GHS08
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH54000
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation