Each bag contains a pre-weighed amount of granulated media, double-bagged and irradiated. For just in time enrichment - add sterile water and sample material.
- Space saving, no bottles taking up shelf space
- Long shelf life
- No autoclaving and optimum solubility
- Dust-free work conditions
The following media are available for the bag system:
Buffered peptone water (BPW), in compliance with ISO 6579-1:2017, is a non selective pre-enrichment broth to increase recovery of bacteria, particularly pathogenic Enterobacteriaceae from foodstuffs
Tryptic soy broth (TSB) used mainly for Media Fill test
Fraser broth base used for the selective enrichment of Listeria spp.
Lactose broth used as a preliminary test for coliform bacteria detection
MacConkey broth for the detection and enumeration of coliforms
Non selective pre-enrichment broth to increase recovery of bacteria, particularly pathogenic Enterobacteriaceae from foodstuffs. The culture medium complies with ISO 6579:2002.
The lab-blender bag contains 5,75 g of BPW to which 225 ml of sterile water and 25 g (or 25 ml) of sample material is added.
More media are available on request.
The following media are approved for the bag system: