Corning® Human recombinant Interleukin-2 (from E. coli)
Supplier: Corning
Certificates
About this item
Interleukin-2 (IL-2), which is also known as T-cell growth factor, plays a central role in cell-mediated immune response.
IL-2 is produced by the helper/inducer subset of T-cells when activated by mitogens such as PHA, or by interaction with antigen/MHC complexes on antigen-presenting cells.
Tested for ability to stimulate proliferation of the mouse T-cell line CTLL. Filtered (0.2 µm membrane) and tested and found negative for the presence of bacteria, fungi, and mycoplasma. Tested for endotoxin (LAL assay).
It was first identified by its ability to promote long-term proliferation of activated T-cells.
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Environmentally Preferable:Low Manufacturing Impact
- Storage conditions:Stable for at least three months at −70 °C (lyophilised) or one month at −70 °C (solubilised). Avoid multiple freeze-thaw cycles.
- Environmental certification:ISO 14001 - Bedford
- Protein/peptide name:IL-2
- Purity:>96% by SDS-PAGE
Tested for ability to stimulate proliferation of the mouse T-cell line CTLL
Filtered (0,2 µm membrane) and tested and found negative for bacteria, fungi and mycoplasma
Tested for endotoxin (LAL assay) - Molecular weight:15,5 kD
- Concentration:10 BRMP units/ml of medium depending on cell type
- Formulation:Frozen in 55 mM sodium phosphate buffer, containing 53 mM sucrose and 0,4% HAS
- Protein synonyms:Interleukin-2
Specifications
Frequently Bought Together