VWR®, Bottles, Wide Neck, HDPE, Rectangular or Square, with Screw Cap
Leverandør: VWR Collection
Sertifikat
Om dette produktet
HDPE.
- Good chemical resistance
- Heat resistant up to +120 °C
- Leakproof
For collecting, storing and shipping samples, or storing liquids and dry substances. Space saving design with rectangular or square base. 2 and 4 litre bottles have integrated handle design.