Bottles, wide neck, with screw cap
Supplier: BURKLE
Certificates
About this item
HDPE, translucent, with PP screw cap.
- Embossed contents scale
- Absolutely leakproof
- Suitable for use with foodstuffs
Bottle for universal use. The medium-sized bottle opening ensures good filling and safe pouring.