Tubing clamps, swing jaw, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Zinc plated steel. Swing jaw clamps for use with all flexible tubing.
- Attach at any point on tubing without disconnecting tubing
- Zinc plated steel is chemical and corrosion resistant
- Knurled nut applies clamping force through smooth clamping surfaces
- Autoclavable