Order Entry
Norway
ContactUsLinkComponent
Cell culture system, Corning® CELLine™ 1000
Cell culture system, Corning® CELLine™ 1000
Katalog # 734-0389
Supplier:  Corning
undefined
Cell culture system, Corning® CELLine™ 1000
Katalog # 734-0389
Supplier:  Corning
Specifications
  • Recommended working volume:
    1015 ml
  • Description:
    CELLine™ 1000
  • Pk:
    3
Specifications
About this item
CELLine™ 1000, a membrane-based, disposable cell cultivation system, guarantees high cell densities and is easy to use for recombinant protein expression and high yield monoclonal antibody (mAb) production.
  • The CELLine™ system yields antibody concentrations comparable to that of ascites - one CELLine™ flask can be used to produce as much antibody as 12 mice
  • Harvest volumes result in antibody concentrations that are 50 to 100 times higher than both roller bottles and tissue culture flasks