Order Entry
Norway
Orders LinkContactUsLinkComponent
 

2,2'-Bithiophene 97%

2,2'-Bithiophene 97%
 
2,2'-Bithiophene 97%
2,2'-Bithiophene 97%

 

 
 :  2,2′-Dithienyl, 2,2′-Bithienyl
  •  :492-97-7
  •  :C₈H₆S₂
  •  :MFCD00005414
  •  :166.27 g/mol
  •  :30…35 °C
  •  :>110 °C
  •  :Refrigerator
  •  :207-767-2

 

 

 

  • Appearance:
    Off-white to dark green to blue Crystals or low melting crystalline mass
  • Infrared spectrum:
    Conforms
  • Melting point:
    30 to 35 °C
  • GC:
    ≥96.0 %
  • Specific optical rotation:
    +42° to +48° (20 °C, 589 nm) (c=3, H2O)
Product Family Options

 

  • Pack type
    Pk
     
     
  • Glass bottle
    2,5 g
       
       
    1 * 2,5 g
    1 * 2,5 g - kr 465,00
    kr 465,00
    1 * 2,5 g
       kr 465,00

  • Glass bottle
    20 g
       
       
    1 * 20 g
    1 * 20 g - kr 2 200,00
    kr 2 200,00
    1 * 20 g
       kr 2 200,00