Order Entry
Norway
ContactUsLinkComponent
 
Dextran, M.W. ~ 70,000
Catalog # MOLE15795395-100G
Supplier:  Molekula
CAS Number:  
Dextran, M.W. ~ 70,000
Catalog # MOLE15795395-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
EINECS:232-677-5
CAS Number:9004-54-0
Formula:(C₆H₁₀O₅)n
MDL Number:MFCD00130935