Test strips, pH, Reflectoquant®, Supelco®
Supplier: Merck
Certificates
About this item
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
- Fast analysis with reliable results
- Barcode calibration
- Low analysis costs
- Environmentally friendly
Specifications
- M-Clarity quality level:MQ100