0
kr 0,00
Order Entry
Logg inn
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Hjem
Bottles
Wash Bottles
Wash bottles, narrow neck, with screw cap, BOLA
Wash bottles, narrow neck, with screw cap, BOLA
:
Bohlender
Wash bottles, narrow neck, with screw cap, BOLA
:
Bohlender
PFA, translucent.
Temperature resistance: from −200 to +250 °C
Universal chemical resistance
Non porous
Graduated