Lithium aluminum hydride 2.4 mol/l in THF, AcroSeal®
:16853-85-3
:
:H₄AlLi
:MFCD00011075
:37.95 g/mol
:–17 °C
:240-877-9
:1411
:4.3,I
:GHS02
:GHS05
:GHS07
:GHS08
- Appearance:Colorless to grey Solution (May develop some turbidity or precipitate)
- Strength:9.5 to 10.5 % (as LiAlH4)