Pyridine
Catalog # PROOCHE126
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2 mL
- Seq:2145
Specifications
About this item
:
Merck Index:13,08060
Boiling Pt:115 °C (1013 hPa)
Melting Pt:–44 °C
Density:0.983 g/cm³ (20 °C)
Flash Pt:17 °C
Storage Temperature:Ambient
EINECS:203-809-9
UN:1282
ADR:3,II
:79.1 g/mol
:GHS02
:GHS07
CAS Number:110-86-1
Formula:C₅H₅N
MDL Number:MFCD00011732