Order Entry
Norway
ContactUsLinkComponent
 
Pyridine ≥99%, GPR RECTAPUR®
  8226.2500
 :  
undefined
Pyridine ≥99%, GPR RECTAPUR®
  8226.2500
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

 

 

  • Assay:
    Min. 99 %
  • Evaporation residue:
    Max. 20 ppm
  • Water:
    Max. 0.5 %

 

 

 :
 :13,08060
 :115 °C (1013 hPa)
 :–44 °C
 :0.983 g/cm³ (20 °C)
 :17 °C
 :Ambient
 :203-809-9
 :1282
 :3,II
 :79.1 g/mol
 :GHS02
 :GHS07
 :110-86-1
 :C₅H₅N
 :MFCD00011732