
2,3-Naphthalenedicarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2169-87-1
:
Formula:C₁₂H₈O₄
MDL Number:MFCD00004104
:216.19 g/mol
Boiling Pt:422 °C (1013 hPa)
Melting Pt:260…262 °C
Storage Temperature:Ambient
EINECS:218-517-7
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:241.0 to 250.0 deg-C