Order Entry
Norway
Orders LinkContactUsLinkComponent
Diphenyl ditelluride 97%

Diphenyl ditelluride 97%

Diphenyl ditelluride 97%
 
Diphenyl ditelluride 97%
Diphenyl ditelluride 97%

 

 
  •  :32294-60-3
  •  :C₁₂H₁₀Te₂
  •  :MFCD00192106
  •  :409.41 g/mol
  •  :64…67 °C
  •  :Ambient
  •  :250-982-1
  •  :3284
  •  :6.1,III

Documents

 

 

  • Appearance:
    Orange to brown Powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    64 to 67 °C
  • HPLC:
    ≥96.0 %

 

Product Family Options

 

  • Pack type
    Pk
     
     
  • Glass bottle for solids
    1 g
       
       
    1 * 1 g
    1 * 1 g - kr 760,00
    kr 760,00
    1 * 1 g
       kr 760,00

     
     
    Specifications
    Cat. No.
    ACRO301840010
  • Glass bottle
    5 g
       
       
    1 * 5 g
    1 * 5 g - kr 2 400,00
    kr 2 400,00
    1 * 5 g
       kr 2 400,00

     
     
    Specifications
    Cat. No.
    ACRO301840050
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.